CO1H324A5DFB CombiPhos Catalysts Inc

3-(Pyridin-2-yl)-5-bromophenylboronic acid

  • Cas Number : 2225175-47-1
  • Molecular Weight : 277.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart