CO1H324A5DFC CombiPhos Catalysts Inc

3-(Pyridin-2-yl)-5-chlorophenylboronic acid

  • Cas Number : 2118958-67-9
  • Molecular Weight : 233.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart