CO1H324A5E06 CombiPhos Catalysts Inc

2-(n-Propoxy)pyrimidine-5-boronic acid

  • Cas Number : 2225179-85-9
  • Molecular Weight : 181.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart