CO1H324A5E11 CombiPhos Catalysts Inc

(2-(2-hydroxypropan-2-yl)pyrimidin-5-yl)boronic acid

  • Cas Number : 1598437-05-8
  • Molecular Weight : 181.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart