CO1H324A5E1D CombiPhos Catalysts Inc

(2-(thiazol-4-yl)pyrimidin-5-yl)boronic acid

  • Cas Number : 2225177-27-3
  • Molecular Weight : 207.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart