CO1H324A5E1F CombiPhos Catalysts Inc

(2-(piperazin-1-yl)pyrimidin-5-yl)boronic acid

  • Cas Number : 1176001-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart