CO1H324A5E21 CombiPhos Catalysts Inc

4-Hydroxypyrimidine-5-boronic acid

  • Cas Number : 2225176-51-0
  • Molecular Weight : 139.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart