CO1H324A5E27 CombiPhos Catalysts Inc

2-Hydroxy-4-ethylpyrimidine-5-boronic acid

  • Cas Number : 2225155-17-7
  • Molecular Weight : 167.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart