CO1H324A5E28 CombiPhos Catalysts Inc

2-Hydroxy-4-(iso-propyl)pyrimidine-5-boronic acid

  • Cas Number : 2225170-78-3
  • Molecular Weight : 181.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart