CO1H324A5E2D CombiPhos Catalysts Inc

2-Chloro-4-(cyclopropyl)pyrimidine-5-boronic acid

  • Cas Number : 2225175-22-2
  • Molecular Weight : 198.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart