CO1H324A5E2E CombiPhos Catalysts Inc

2-Bromo-4-(cyclopropyl)pyrimidine-5-boronic acid

  • Cas Number : 2225176-67-8
  • Molecular Weight : 242.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart