CO1H324A5E36 CombiPhos Catalysts Inc

4-Methyl-2-(ethoxy)pyrimidine-5-boronic acid

  • Cas Number : 1453218-81-9
  • Molecular Weight : 181.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart