CO1H324A5E3D CombiPhos Catalysts Inc

4-Methyl-2-(1H-pyrrol-1-yl)pyrimidine-5-boronic acid

  • Cas Number : 2225181-40-6
  • Molecular Weight : 203.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart