CO1H324A5E3E CombiPhos Catalysts Inc

4-Methyl-2-piperidinopyrimidine-5-boronic acid

  • Cas Number : 2225177-09-1
  • Molecular Weight : 221.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart