CO1H324A5E3F CombiPhos Catalysts Inc

4-Methyl-2-morpholinopyrimidine-5-boronic acid

  • Cas Number : 2077990-06-6
  • Molecular Weight : 223.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart