CO1H324A5E47 CombiPhos Catalysts Inc

4-Methyl-2-(3-methylpiperidino)pyrimidine-5-boronic acid

  • Cas Number : 2225152-35-0
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart