CO1H324A5E4A CombiPhos Catalysts Inc

4-Methyl-2-(4-methylpiperidino)pyrimidine-5-boronic acid

  • Cas Number : 2225175-46-0
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart