CO1H324A5E55 CombiPhos Catalysts Inc

(2-(2-aminopropan-2-yl)pyridin-4-yl)boronic acid

  • Cas Number : 2225181-41-7
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart