CO1H324A5E57 CombiPhos Catalysts Inc

(5-(2-aminopropan-2-yl)pyridin-3-yl)boronic acid

  • Cas Number : 2225177-33-1
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart