CO1H324A5E6D CombiPhos Catalysts Inc

2-Fluoro-6-(3-furyl)pyridine-3-boronic acid

  • Cas Number : 2225154-62-9
  • Molecular Weight : 206.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart