CO1H324A5E72 CombiPhos Catalysts Inc

6-Fluoro-5-trifluoromethylpyridine-3-boronic acid

  • Cas Number : 2225175-78-8
  • Molecular Weight : 208.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart