CO1H324A5E7F CombiPhos Catalysts Inc

2-Methyl-6-(piperidin-3-yl)pyridine-3-boronic acid

  • Cas Number : 2225151-92-6
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart