CO1H324A5E88 CombiPhos Catalysts Inc

(5-CHLORO-4-METHYLPYRIDIN-3-YL)BORONIC ACID

  • Cas Number : 2225172-97-2
  • Molecular Weight : 171.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart