CO1H324A5E89 CombiPhos Catalysts Inc

4-Fluoro-3-bromopyridine-5-boronic acid

  • Cas Number : 2225177-26-2
  • Molecular Weight : 219.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart