CO1H324A5E8B CombiPhos Catalysts Inc

6-Chloropyrazine-2-boronic acid

  • Cas Number : 1310404-64-8
  • Molecular Weight : 158.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart