CO1H324A5E93 CombiPhos Catalysts Inc

3-(Cyclopentyl)pyridine-4-boronic acid

  • Cas Number : 2225180-02-7
  • Molecular Weight : 191.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart