CO1H324A5E98 CombiPhos Catalysts Inc

5-Methyl-4-methoxypyridine-3-boronic acid

  • Cas Number : 1416423-91-0
  • Molecular Weight : 166.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart