CO1H324A5EBA CombiPhos Catalysts Inc

(2-propylpyridin-4-yl)boronic acid

  • Cas Number : 2225180-61-8
  • Molecular Weight : 165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart