CO1H324A5ECC CombiPhos Catalysts Inc

2-(Dimethylamino)pyridine-4-boronic acid

  • Cas Number : 1207749-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart