CO1H324A5ECE CombiPhos Catalysts Inc

2-(Methylethylamino)pyridine-4-boronic acid

  • Cas Number : 2225152-03-2
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart