CO1H324A5ED4 CombiPhos Catalysts Inc

2-(3-Methyl-1H-pyrazol-1-yl)pyridine-4-boronic acid

  • Cas Number : 2225172-82-5
  • Molecular Weight : 203.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart