CO1H324A5EDA CombiPhos Catalysts Inc

2-(3-Methylpiperidin-1-yl)pyridine-4-boronic acid

  • Cas Number : 2225152-63-4
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart