CO1H324A5EDB CombiPhos Catalysts Inc

2-(4-Methylpiperidin-1-yl)pyridine-4-boronic acid

  • Cas Number : 1704063-45-5
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart