CO1H324A5EE6 CombiPhos Catalysts Inc

2-(Tetrahydrofuran-3-yloxy)pyridine-4-boronic acid

  • Cas Number : 2225152-13-4
  • Molecular Weight : 209.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart