CO1H324A5EEC CombiPhos Catalysts Inc

(2-ethylpyridin-4-yl)boronic acid

  • Cas Number : 1189545-99-0
  • Molecular Weight : 150.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart