CO1H324A5EF0 CombiPhos Catalysts Inc

(2-cyclopentylpyridin-4-yl)boronic acid

  • Cas Number : 2225172-93-8
  • Molecular Weight : 191.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart