CO1H324A5EF4 CombiPhos Catalysts Inc

2-(4-Methoxyphenyl)pyridine-4-boronic acid

  • Cas Number : 912844-93-0
  • Molecular Weight : 229.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart