CO1H324A5EF8 CombiPhos Catalysts Inc

2-(3,5-Difluorophenyl)pyridine-4-boronic acid

  • Cas Number : 2225180-11-8
  • Molecular Weight : 235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart