CO1H324A5EFB CombiPhos Catalysts Inc

(2-(thiophen-2-yl)pyridin-4-yl)boronic acid

  • Cas Number : 2096336-59-1
  • Molecular Weight : 205.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart