CO1H324A5EFF CombiPhos Catalysts Inc

2-(2-Tolyl)pyridine-5-boronic acid

  • Cas Number : 2225177-38-6
  • Molecular Weight : 213.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart