CO1H324A5F02 CombiPhos Catalysts Inc

2-(2-Fluorophenyl)pyridine-5-boronic acid

  • Cas Number : 1072944-20-7
  • Molecular Weight : 217.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart