CO1H324A5F0E CombiPhos Catalysts Inc

2-(1H-Pyrrol-1-yl)pyridine-5-boronic acid

  • Cas Number : 899436-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart