CO1H324A5F11 CombiPhos Catalysts Inc

2-(4-Methylimidazol-1-yl)pyridine-5-boronic acid

  • Cas Number : 2225180-12-9
  • Molecular Weight : 203.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart