CO1H324A5F26 CombiPhos Catalysts Inc

5-(n-Propyl)pyridine-3-boronic acid

  • Cas Number : 2225154-41-4
  • Molecular Weight : 165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-propyl-3-pyridinylboronic acid
Inchi Code 1S/C8H12BNO2/c1-2-3-7-4-8(9(11)12)6-10-5-7/h4-6,11-12H,2-3H2,1H3
Inchi Key DONIINMODBXJPH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart