CO1H324A5F2A CombiPhos Catalysts Inc

(5-cyclopentylpyridin-3-yl)boronic acid

  • Cas Number : 2225178-81-2
  • Molecular Weight : 191.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart