CO1H324A5F2F CombiPhos Catalysts Inc

5-(2-Tolyl)pyridine-3-boronic acid

  • Cas Number : 1104125-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart