CO1H324A5F30 CombiPhos Catalysts Inc

5-(3-Tolyl)pyridine-3-boronic acid

  • Cas Number : 1714084-47-5
  • Molecular Weight : 213.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart