CO1H324A5F39 CombiPhos Catalysts Inc

5-(4-Trifluoromethylphenyl)pyridine-3-boronic acid

  • Cas Number : 1237739-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart