CO1H324A5F47 CombiPhos Catalysts Inc

(5-(furan-3-yl)pyridin-3-yl)boronic acid

  • Cas Number : 2225154-44-7
  • Molecular Weight : 188.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart