CO1H324A5F4F CombiPhos Catalysts Inc

5-(4-Methyl-1H-pyrazol-1-yl)pyridine-3-boronic acid

  • Cas Number : 2225156-02-3
  • Molecular Weight : 203.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart